Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18159

Sigma-Aldrich

(R)-(+)-4-Methoxy-α-methylbenzylamine

≥95.0% (GC)

Synonym(s):

(R)-(+)-1-(4-Methoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Beilstein/REAXYS Number:
2413029
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

assay

≥95.0% (GC)

optical activity

[α]20/D +32±2°, neat

optical purity

enantiomeric ratio: ≥99.0:1.0 (GC)

refractive index

n20/D 1.533

density

1.024 g/mL at 20 °C (lit.)

SMILES string

COc1ccc(cc1)[C@@H](C)N

InChI

1S/C9H13NO/c1-7(10)8-3-5-9(11-2)6-4-8/h3-7H,10H2,1-2H3/t7-/m1/s1

Inchi Key

JTDGKQNNPKXKII-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service