Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177334

Sigma-Aldrich

Methylglyoxal solution

40 wt. % in H2O

Synonym(s):

Acetylformaldehyde, Pyruvaldehyde, Pyruvic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCHO
CAS Number:
Molecular Weight:
72.06
Beilstein/REAXYS Number:
906750
MDL number:
UNSPSC Code:
12352103

concentration

40 wt. % in H2O

refractive index

n20/D 1.4209

density

1.178 g/mL at 25 °C

SMILES string

[H]C(=O)C(C)=O

InChI

1S/C3H4O2/c1-3(5)2-4/h2H,1H3

InChI key

AIJULSRZWUXGPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service