Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

176869

Sigma-Aldrich

Trimethylamine N-oxide dihydrate

98%

Synonym(s):

TMANO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3NO · 2H2O
CAS Number:
Molecular Weight:
111.14
Beilstein/REAXYS Number:
3612927
EC Number:
MDL number:
UNSPSC Code:
12352000

assay

98%

mp

95-99 °C (lit.)

SMILES string

O.O.C[N+](C)(C)[O-]

InChI

1S/C3H9NO.2H2O/c1-4(2,3)5;;/h1-3H3;2*1H2

InChI key

PGFPZGKEDZGJQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • C-H bond cleavage
  • Oxidation reactions (oxidant)

  • Decarbonylating agent for solvent-free reactions

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Wangler et al.
Physical chemistry chemical physics : PCCP, 20(16), 11317-11326 (2018-04-12)
This work presents an approach that expresses the Michaelis constant KaM and the equilibrium constant Kth of an enzymatic peptide hydrolysis based on thermodynamic activities instead of concentrations. This provides KaM and Kth values that are independent of any co-solvent.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service