Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

176699

Styrene-d8

≥98 atom % D

Synonym(s):

Octadeuterostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD=CD2
CAS Number:
Molecular Weight:
112.20
Beilstein/REAXYS Number:
2327045
MDL number:
Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

refractive index

n20/D 1.5445 (lit.)

bp

145-146 °C (lit.)

mp

-31 °C

density

0.979 g/mL at 25 °C (lit.)

mass shift

M+8

SMILES string

[2H]\C([2H])=C(/[2H])c1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2/i1D2,2D,3D,4D,5D,6D,7D

InChI key

PPBRXRYQALVLMV-GDALLCCDSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service