Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

173118

Sigma-Aldrich

4-Vinyl-1-cyclohexene

97%

Synonym(s):

4-Ethenyl-1-cyclohexene, NSC 15760

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CH=CH2
CAS Number:
Molecular Weight:
108.18
Beilstein/REAXYS Number:
1901553
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

3.76 (vs air)

vapor pressure

10.2 mmHg ( 25 °C)

assay

97%

autoignition temp.

517 °F

refractive index

n20/D 1.463 (lit.)

bp

126-127 °C (lit.)

mp

−101 °C (lit.)

density

0.831 g/mL at 20 °C
0.832 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1CCC=CC1

InChI

1S/C8H12/c1-2-8-6-4-3-5-7-8/h2-4,8H,1,5-7H2

InChI key

BBDKZWKEPDTENS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service