Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

168130

Sigma-Aldrich

2,4-Quinolinediol monosodium salt hydrate

90% (Purity measured on dry basis), technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6NNaO2 · xH2O
CAS Number:
Molecular Weight:
183.14 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

assay

90% (Purity measured on dry basis)

form

powder

impurities

30% water

SMILES string

[Na+].[H]O[H].Oc1cc([O-])c2ccccc2n1

InChI

1S/C9H7NO2.Na.H2O/c11-8-5-9(12)10-7-4-2-1-3-6(7)8;;/h1-5H,(H2,10,11,12);;1H2/q;+1;/p-1

InChI key

TUOHINFEBZWDSZ-UHFFFAOYSA-M

Application

2,4-Quinolinediol was used in the synthesis of novel hetarylazoquinoline dyes.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and investigations of the absorption spectra of hetarylazo disperse dyes derived from 2, 4-quinolinediol.
Sener L, et al.
Dyes and Pigments, 70(2), 143-148 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service