Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

166995

Sigma-Aldrich

Ethyl cyclohexanone-2-carboxylate

98%

Synonym(s):

Ethyl 2-oxocyclohexanecarboxylate, 2-(Ethoxycarbonyl)cyclohexanone, Ethyl-2-oxocyclohexancarboxylat

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C6H9CO2C2H5
CAS Number:
Molecular Weight:
170.21
Beilstein/REAXYS Number:
608356
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

assay

98%

form

liquid

availability

available only in EU

refractive index

n20/D 1.477 (lit.)

bp

106 °C/11 mmHg (lit.)

density

1.064 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)C1CCCCC1=O

InChI

1S/C9H14O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h7H,2-6H2,1H3

InChI key

FGSGHBPKHFDJOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service