Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16548

Sigma-Aldrich

4,4,5,5,6,6,7,7,8,8,9,9,9-Tridecafluorononyl iodide

≥97% (GC)

Synonym(s):

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-9-iodononane, 1H,1H,2H,2H,3H,3H-Perfluorononyl iodide, 3-(Perfluorohexyl)propyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6F13I
CAS Number:
Molecular Weight:
488.03
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥97% (GC)

form

liquid

refractive index

n20/D 1.369

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCI

InChI

1S/C9H6F13I/c10-4(11,2-1-3-23)5(12,13)6(14,15)7(16,17)8(18,19)9(20,21)22/h1-3H2

InChI key

DBWLHXGWJWZTDX-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service