163503
α,α-Dimethyl-β-methylsuccinimide
99%
Synonym(s):
2,2,3-Trimethylsuccinimide
About This Item
Recommended Products
assay
99%
form
solid
mp
119-121 °C (lit.)
SMILES string
CC1C(=O)NC(=O)C1(C)C
InChI
1S/C7H11NO2/c1-4-5(9)8-6(10)7(4,2)3/h4H,1-3H3,(H,8,9,10)
Inchi Key
SUPYQTKKYLUOBW-UHFFFAOYSA-N
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service