Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16303

Sigma-Aldrich

4′-Bromobenzo-15-crown-5

≥97.0%

Synonym(s):

(Benzo-15-crown-5)-4′-yl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19BrO5
CAS Number:
Molecular Weight:
347.20
Beilstein/REAXYS Number:
1587587
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

assay

≥97.0%

mp

68-71 °C

SMILES string

Brc1ccc2OCCOCCOCCOCCOc2c1

Other Notes

Starting material for preparing substituted benzo-15-crown-5 derivatives[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Cefelin et al.
Collection of Czechoslovak Chemical Communications, 57, 472-472 (1992)
T. Okano et al.
Journal of Organometallic Chemistry, 346, 267-267 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service