Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

161764

Sigma-Aldrich

1,4-Dithioerythritol

≥99%

Synonym(s):

erythro-1,4-Dimercapto-2,3-butanediol, erythro-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein/REAXYS Number:
1719756
EC Number:
MDL number:
UNSPSC Code:
12352201

assay

≥99%

mp

82-84 °C (lit.)

SMILES string

O[C@@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for disulfide groups.[1] Used to prepare a matrix for fast-atom bombardment mass spectroscopy.[2]
Reagent for maintaining −SH groups in the reduced state; quantitatively reduces disulfides.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid Communications in Mass Spectrometry, 5, 469-469 (1991)
Aldrichimica Acta, 4, 33-33 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service