Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

154318

Sigma-Aldrich

3,4-Dihydroxy-L-phenylalanine

optical purity ee: 99% (GLC), 99%

Synonym(s):

3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.19
Beilstein/REAXYS Number:
2215169
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

optical activity

[α]25/D −11°, c = 1 in 1 M HCl

optical purity

ee: 99% (GLC)

mp

276-278 °C (lit.)

SMILES string

N[C@@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

WTDRDQBEARUVNC-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molly J Crockett et al.
Current biology : CB, 25(14), 1852-1859 (2015-07-07)
An aversion to harming others is a core component of human morality and is disturbed in antisocial behavior. Deficient harm aversion may underlie instrumental and reactive aggression, which both feature in psychopathy. Past work has highlighted monoaminergic influences on aggression
A P de Jong et al.
Clinica chimica acta; international journal of clinical chemistry, 171(1), 49-61 (1988-01-15)
L-3-(3,4-Dihydroxyphenyl)alanine (DOPA) and its 3-O-methyl metabolite (OMD) were measured in plasma and cerebrospinal fluid by a new assay which combines N,O-acetylation of amino acids in aqueous media, preparation of pentafluorobenzyl esters under anhydrous conditions, and analysis by gas chromatography-electron capture
Damian M Herz et al.
Annals of neurology, 75(6), 829-836 (2014-06-04)
In Parkinson disease (PD), long-term treatment with the dopamine precursor levodopa gradually induces involuntary "dyskinesia" movements. The neural mechanisms underlying the emergence of levodopa-induced dyskinesias in vivo are still poorly understood. Here, we applied functional magnetic resonance imaging (fMRI) to
Nicolas Morin et al.
Experimental neurology, 256, 105-116 (2013-01-31)
The treatment of motor symptoms of Parkinson disease (PD) with the dopamine (DA) precursor, l-3,4-dihydroxyphenylalanine (l-DOPA) introduced 50years ago still remains a very effective medication. However, involuntary movements termed l-DOPA-induced dyskinesias (LID) appear in the vast majority of PD patients
Vincent J J Odekerken et al.
Neurology, 84(13), 1355-1361 (2015-03-01)
To assess the neuropsychological outcome 12 months after bilateral deep brain stimulation (DBS) of the globus pallidus pars interna (GPi) or subthalamic nucleus (STN) for advanced Parkinson disease. We randomly assigned patients to receive either GPi DBS or STN DBS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service