150622
Benzofuroxan
98%
Synonym(s):
Benzofurazan 1-oxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4N2O2
CAS Number:
Molecular Weight:
136.11
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
assay
98%
mp
69-71 °C (lit.)
SMILES string
[O-][n+]1onc2ccccc12
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service