Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150568

Sigma-Aldrich

(−)-Glutathione, oxidized

Synonym(s):

L-Glutathione oxidized, GSSG, Glutathiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HO2CCH(NH2)CH2CH2CONHCH(CONHCH2CO2H)CH2S-]2
CAS Number:
Molecular Weight:
612.63
Beilstein/REAXYS Number:
1718700
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

optical activity

[α]20/D -92.0 to -106.0°, c = 2% in H2O

impurities

≤6% water (Karl Fischer)

mp

178 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[H][C@](N)(CCC(=O)N[C@@]([H])(CSSC[C@]([H])(NC(=O)CC[C@]([H])(N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C20H32N6O12S2/c21-9(19(35)36)1-3-13(27)25-11(17(33)23-5-15(29)30)7-39-40-8-12(18(34)24-6-16(31)32)26-14(28)4-2-10(22)20(37)38/h9-12H,1-8,21-22H2,(H,23,33)(H,24,34)(H,25,27)(H,26,28)(H,29,30)(H,31,32)(H,35,36)(H,37,38)/t9-,10-,11-,12-/m0/s1

InChI key

YPZRWBKMTBYPTK-BJDJZHNGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrogen acceptor in the enzymic determination of NADP and NADPH.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 4144-4144 (1993)
Doina Gherghel et al.
Investigative ophthalmology & visual science, 54(5), 3333-3339 (2013-04-20)
To investigate in parallel the systemic glutathione levels of patients suffering from primary open angle glaucoma (POAG) or normal tension glaucoma (NTG) with comparable functional loss. Thirty-four POAG patients, 30 NTG patients, and 53 controls were subjected to blood analysis
Yan Wang et al.
Environmental toxicology and pharmacology, 35(1), 109-116 (2013-01-01)
Lead is linked to many reproductive problems. This study was to explore the chronic effects of low lead level on expressions of Nrf2 and Mrp1 in rats' testes. Maternal SD rats were administered lead acetate from 10 days before gestation
Chi-Chang Huang et al.
Molecules (Basel, Switzerland), 18(2), 1741-1754 (2013-02-26)
Several studies have been shown that accelerated apoptosis is involved in post-exercise lymphocytopenia and tissue damage after high-intensity exercise. Ganoderma tsugae (GT) is one of the well-known medicinal mushrooms that possess various pharmacological functions. This mushroom has traditionally been used
Redox control: A black hole for oxidized glutathione.
Jakob R Winther et al.
Nature chemical biology, 9(2), 69-70 (2013-01-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service