15054
Boc-Aad-OH
≥98.0% (TLC)
Synonym(s):
Boc-L-homoglutamic acid, N-Boc-L-2-aminoadipic acid
About This Item
assay
≥98.0% (TLC)
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)N[C@@H](CCCC(O)=O)C(O)=O
InChI
1S/C11H19NO6/c1-11(2,3)18-10(17)12-7(9(15)16)5-4-6-8(13)14/h7H,4-6H2,1-3H3,(H,12,17)(H,13,14)(H,15,16)/t7-/m0/s1
InChI key
QDTDLMJRZPSKDM-ZETCQYMHSA-N
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service