Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150517

Sigma-Aldrich

Perhydrofluorene

97%

Synonym(s):

Dodecahydrofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22
CAS Number:
Molecular Weight:
178.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.501 (lit.)

bp

253 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

C1CCC2C(C1)CC3CCCCC23

InChI

1S/C13H22/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h10-13H,1-9H2

InChI key

OLWAZOBRCQWWDB-UHFFFAOYSA-N

General description

The thermal decomposition of perhydrofluorene (dodecahydrofluorene) was studied to determine the formation of aromatics from naphthenes[1].

Application

Perhydrofluorene (Dodecahydrofluorene) was used as a hydrogen storage compound[2].

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

147.2 °F - closed cup

flash_point_c

64 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zimmermann G, et al.
Journal of Analytical and Applied Pyrolysis, 7(3), 195-205 (1985)
Exploring the Fuel Limits of Direct Oxidation Proton Exchange Membrane Fuel Cells with Platinum Based Electrocatalysts.
Journal of the Electrochemical Society, 159(4), B371-B377 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service