Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

149462

Sigma-Aldrich

Diethyl phenylphosphonite

97%

Synonym(s):

Diethoxyphenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P(OC2H5)2
CAS Number:
Molecular Weight:
198.20
EC Number:
MDL number:
UNSPSC Code:
12350000

assay

97%

refractive index

n20/D 1.51 (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

CCOP(OCC)c1ccccc1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service