Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

149373

Sigma-Aldrich

Bicyclo[4.3.0]nona-3,6(1)-diene

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12
CAS Number:
Molecular Weight:
120.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

assay

85%

refractive index

n20/D 1.508 (lit.)

bp

178 °C (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

C1CC2=C(C1)CC=CC2

Other Notes

remainder indan

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

50.0 °F - closed cup

flash_point_c

10 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service