Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

148059

Sigma-Aldrich

4-tert-Butylphenyl glycidyl ether

98%

Synonym(s):

4-tert-Butylphenyl 2,3-epoxypropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.515 (lit.)

bp

165-170 °C/14 mmHg (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(OCC2CO2)cc1

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

289.9 °F

flash_point_c

143.3 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service