Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

145416

Sigma-Aldrich

2,6-Diiodo-4-nitrophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I2C6H2(NO2)OH
CAS Number:
Molecular Weight:
390.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

152-154 °C (lit.)

SMILES string

Oc1c(I)cc(cc1I)[N+]([O-])=O

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Daengsvang
The Southeast Asian journal of tropical medicine and public health, 11(3), 359-362 (1980-09-01)
Twelve doses of Ancylol Disophenol at 0.05 ml per pound body weight administered subcutaneously was shown to be an effective chemotherapeutic agent against migrating stage of G. spinigerum in cats. A further investigation is required to determine whether this regimen
Occurrence of resistance to anthelmintics in sheep in Paraná State, Brazil.
V T Soccol et al.
The Veterinary record, 139(17), 421-422 (1996-10-26)
C A Hall et al.
Research in veterinary science, 31(1), 104-106 (1981-07-01)
Two long-acting anthelmintics, closantel and disophenol, were tested by treatment 90, 60 and 30 days before challenge with a thiabendazole selected laboratory strain of Haemonchus contortus. The sheep were slaughtered 28 days after infection for total worm counts. A significant
G W Hutchinson
Research in veterinary science, 30(2), 175-180 (1981-03-01)
Haematological parameters and liver specific serum enzymes were examined in pigs during the first 12 weeks of liver migration of larvae following experimental infection with 1000 infective Stephanurus dentatus larvae. No significant changes in total red blood cell counts, packed
R K Reinecke et al.
The Onderstepoort journal of veterinary research, 51(1), 71-79 (1984-03-01)
Sheep grazed on natural pastures heavily infested with infective larvae of Haemonchus contortus. Sixty-eight weaned Merinos were divided into 6 groups on Day 0 (23 November 1977), and on Day +14 (7 December) 79 Merinos were divided into 7 groups.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service