Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14534

Sigma-Aldrich

1,3-Bis(3-aminopropyl)-1,1,3,3-tetramethyldisiloxane

≥95.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H28N2OSi2
CAS Number:
Molecular Weight:
248.51
Beilstein/REAXYS Number:
1760876
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥95.0% (T)

refractive index

n20/D 1.451

bp

132 °C/11 mmHg (lit.)

density

0.901 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(CCCN)O[Si](C)(C)CCCN

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service