Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

145033

Sigma-Aldrich

3,4-Epoxytetrahydrothiophene-1,1-dioxide

technical grade

Synonym(s):

3,4-Epoxysulfolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3S
CAS Number:
Molecular Weight:
134.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

mp

145-150 °C (lit.)

SMILES string

O=S1(=O)CC2OC2C1

InChI

1S/C4H6O3S/c5-8(6)1-3-4(2-8)7-3/h3-4H,1-2H2

InChI key

SZAIAWVGWTXVMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service