Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14401

Sigma-Aldrich

Zinc

≥99%, powder (coarse)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

1 mmHg ( 487 °C)

assay

≥99%

form

powder (coarse)

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

cation traces

As: ≤0.2 mg/kg
Cd: ≤50 mg/kg
Fe: ≤50 mg/kg
Pb: ≤100 mg/kg
Sn: ≤10 mg/kg

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

application

Used as reducing agent; used for preparation of organozinc reagents; used as catalyst for cross coupling

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kise, N.; Ueda, N. et al
Tetrahedron Letters, 42, 2365-2365 (2001)
Paul, S.; Nanda, P.; Gupta, R.; Loupy, A.
Synthesis, 2872-2872 (2003)
Neo, A. G.; Delgado, J. et al.
Tetrahedron Letters, 46, 23-23 (2005)
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service