Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

142522

Sigma-Aldrich

2-Benzylphenol

99%

Synonym(s):

α-Phenyl-o-cresol, 2-Hydroxydiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H4OH
CAS Number:
Molecular Weight:
184.23
Beilstein/REAXYS Number:
2045725
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

bp

312 °C (lit.)

mp

49-51 °C (lit.)

SMILES string

Oc1ccccc1Cc2ccccc2

InChI

1S/C13H12O/c14-13-9-5-4-8-12(13)10-11-6-2-1-3-7-11/h1-9,14H,10H2

InChI key

CDMGNVWZXRKJNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service