Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

142093

Sigma-Aldrich

N,N-Dichlorourethane

technical grade, 90%

Synonym(s):

DCU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2NCO2C2H5
CAS Number:
Molecular Weight:
157.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

assay

90%

refractive index

n20/D 1.46 (lit.)

bp

55-56 °C/15 mmHg (lit.)

density

1.349 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)N(Cl)Cl

InChI

1S/C3H5Cl2NO2/c1-2-8-3(7)6(4)5/h2H2,1H3

InChI key

PCGGNOFZZFFFLU-UHFFFAOYSA-N

General description

N,N-Dichlorourethane undergoes addition reaction with unsaturated compounds to yield β-chlorocarbamates. It is a pseudohalogen and is an attractive halogenoid reagent. It reacts with indole and indole-2- or 3-carboxylic acid to yield 3,3,5-trichlorooxindole.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

206.6 °F - closed cup

flash_point_c

97 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pseudohalogens. X. Effect of some electronic or steric factors on the addition of N, N-dichlorourethane to unsaturated compounds.
Foglia TA and Swern D.
The Journal of Organic Chemistry, 33(2), 766-771 (1968)
Pseudohalogens. XII. Reaction of N, N-dichlorourethane with indole and derivatives.
Foglia TA and Swern D.
The Journal of Organic Chemistry, 33(12), 4440-4442 (1968)
Rémi Rosière et al.
International journal of oncology, 47(3), 1131-1142 (2015-07-24)
Besides the numerous advantages of a chemotherapy administered by the inhalation route for lung cancer therapy, dry powder for inhalation (DPI) offers many advantages compared to other techniques and seems to be a technique that is well-adapted to an anticancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service