Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139157

Sigma-Aldrich

3-Indolebutyric acid

98%

Synonym(s):

4-(3-Indolyl)butanoic acid, IBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2
CAS Number:
Molecular Weight:
203.24
Beilstein/REAXYS Number:
171120
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

124-125.5 °C (lit.)

SMILES string

OC(=O)CCCc1c[nH]c2ccccc12

InChI

1S/C12H13NO2/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11/h1-2,5-6,8,13H,3-4,7H2,(H,14,15)

InChI key

JTEDVYBZBROSJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emeline L Maillet et al.
Journal of medicinal chemistry, 52(21), 6931-6935 (2009-10-13)
We show that phenoxyauxin herbicides and lipid-lowering fibrates inhibit human but not rodent T1R3. T1R3 as a coreceptor in taste cells responds to sweet compounds and amino acids; in endocrine cells of gut and pancreas T1R3 contributes to glucose sensing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service