All Photos(1)
About This Item
Linear Formula:
FC6H3-4-(OH)CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
199.18
EC Number:
MDL number:
UNSPSC Code:
12352103
assay
98%
mp
280 °C (dec.) (lit.)
SMILES string
NC(Cc1ccc(O)c(F)c1)C(O)=O
InChI
1S/C9H10FNO3/c10-6-3-5(1-2-8(6)12)4-7(11)9(13)14/h1-3,7,12H,4,11H2,(H,13,14)
InChI key
VIIAUOZUUGXERI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service