Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

134414

Sigma-Aldrich

Succinic anhydride

97%

Synonym(s):

Dihydro-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O3
CAS Number:
Molecular Weight:
100.07
Beilstein/REAXYS Number:
108441
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

1 mmHg ( 92 °C)

assay

97%

bp

261 °C (lit.)

mp

118-120 °C (lit.)

SMILES string

O=C1CCC(=O)O1

InChI

1S/C4H4O3/c5-3-1-2-4(6)7-3/h1-2H2

InChI key

RINCXYDBBGOEEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

314.6 °F

flash_point_c

157 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenlian Zhu et al.
Magnetic resonance in medicine, 59(4), 679-685 (2008-02-28)
Pretargeting of receptors is a useful approach in molecular imaging and therapy to reduce background noise or toxicity and enhance selectivity. In this study a three-step pretargeting approach that includes a biotinylated antibody, avidin/streptavidin, and a biotinylated imaging agent is
Christian J Koehler et al.
Analytical chemistry, 83(12), 4775-4781 (2011-05-03)
Recently, we introduced a novel approach for protein quantification based on isobaric peptide termini labeling (IPTL). In IPTL, both peptide termini are dervatized in two separate chemical reactions with complementary isotopically labeled reagents to generate isobaric peptide pairs. Here, we
Peisheng Xu et al.
Biomacromolecules, 11(9), 2352-2358 (2010-08-11)
Zwitterionic chitosan, a chitosan derivative with a unique pH-dependent charge profile, was employed to create a stealth coating on the cationic surface of drug carriers. Zwitterionic chitosans were synthesized by amidation of chitosan with succinic anhydride. The succinic anhydride-conjugated chitosan
Jean-François Poulin et al.
International journal of pharmaceutics, 405(1-2), 47-54 (2010-12-07)
The use of succinylated β-lactoglobulin as a novel functional tablet excipient for the protection of probiotic bacteria against the adverse gastric conditions and their delivery in the intestine was studied. Tablets were produced by direct compression of a dry mixture
Katharine C Payne et al.
Environmental science & technology, 46(14), 7725-7730 (2012-06-26)
Wood-derived cellulosic fibers prepared in different ways were successfully employed to absorb simulated crude oil, demonstrating their possible use as absorbents in the case of oil spills. When dry fibers were used, the highest sorption capacity (six parts of oil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service