Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13401

Sigma-Aldrich

Sodium

≥99%, pieces (stored under mineral oil, heavy)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na
CAS Number:
Molecular Weight:
22.99
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

vapor pressure

1 mmHg ( 440 °C)

assay

≥99%

form

pieces (stored under mineral oil, heavy)

reaction suitability

reagent type: reductant

resistivity

4.69 μΩ-cm, 20°C

bp

883 °C (lit.)

mp

97.8 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

Ca: ≤1000 mg/kg
K: ≤500 mg/kg

SMILES string

[Na]

InChI

1S/Na

InChI key

KEAYESYHFKHZAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supriya S Jalimarada et al.
Investigative ophthalmology & visual science, 54(6), 4330-4340 (2013-06-08)
Mutations in SLC4A11, a member of the SLC4 superfamily of bicarbonate transporters, give rise to corneal endothelial cell dystrophies. SLC4A11 is a putative Na⁺ borate and Na⁺:OH⁻ transporter. Therefore we ask whether SLC4A11 in corneal endothelium transports borate (B[OH]₄⁻), bicarbonate
Nicole Li et al.
American heart journal, 166(5), 815-822 (2013-11-02)
Cardiovascular diseases are the leading cause of death and disability in China. High blood pressure caused by excess intake of dietary sodium is widespread and an effective sodium reduction program has potential to improve cardiovascular health. This study is a
Julijana Vuchkova et al.
American journal of physiology. Renal physiology, 307(5), F525-F532 (2014-07-06)
Albuminuria in nephrotic states is thought to arise from the formation of large pores in the glomerular capillary wall as large hydrodynamic probes, like Ficoll, have increased fractional clearance. In the present study, we tested for large pore formation in
Martin O'Donnell et al.
The New England journal of medicine, 371(7), 612-623 (2014-08-15)
The optimal range of sodium intake for cardiovascular health is controversial. We obtained morning fasting urine samples from 101,945 persons in 17 countries and estimated 24-hour sodium and potassium excretion (used as a surrogate for intake). We examined the association
Etana Padan
Biochimica et biophysica acta, 1837(7), 1047-1062 (2013-12-24)
The crystal structure of down-regulated NhaA crystallized at acidic pH4 [21] has provided the first structural insights into the antiport mechanism and pH regulation of a Na(+)/H(+) antiporter [22]. On the basis of the NhaA crystal structure [21] and experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service