Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

131458

Sigma-Aldrich

Phenylpropargyl aldehyde diethyl acetal

98%

Synonym(s):

Phenylpropiolic aldehyde diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCH(OC2H5)2
CAS Number:
Molecular Weight:
204.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

liquid

refractive index

n20/D 1.517 (lit.)

bp

99-100 °C/2 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(OCC)C#Cc1ccccc1

InChI

1S/C13H16O2/c1-3-14-13(15-4-2)11-10-12-8-6-5-7-9-12/h5-9,13H,3-4H2,1-2H3

InChI key

DTEGZYXCDQFSBZ-UHFFFAOYSA-N

Application

Phenylpropargyl aldehyde diethyl acetal was used in the preparation of Z-cinnamaldehyde[1].

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C H Williams et al.
The Biochemical journal, 256(3), 911-915 (1988-12-15)
3-Amino-1-phenylprop-1-ene (cinnamylamine) and some derivatives were examined as substrates for monoamine oxidases A and B in mitochondria. All of the amines examined were readily oxidized by monoamine oxidase B but much less readily by monoamine oxidase A. E-Cinnamylamine was found

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service