Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

131431

Sigma-Aldrich

DL-Norephedrine hydrochloride

≥99%

Synonym(s):

(±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
CAS Number:
Molecular Weight:
187.67
Beilstein/REAXYS Number:
3914299
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

assay

≥99%

mp

194-196 °C (lit.)

SMILES string

Cl.C[C@@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9-;/m1./s1

InChI key

DYWNLSQWJMTVGJ-PRCZDLBKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mixed-acting sympathomimetic amine.

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuei Hui Chan et al.
Journal of analytical toxicology, 33(3), 162-166 (2009-04-18)
Sho-seiryu-to is one of the most common Traditional Chinese Medicine preparations for the attenuation of colds. Ephedrae Herba is one of the prescriptions of Sho-seiryu-to. The major ingredients of Ephedrae Herba, ephedrines, are banned substances on the World Anti-Doping Agency
Raz Krizevski et al.
Journal of ethnopharmacology, 114(3), 432-438 (2007-10-12)
Khat (Catha edulis Forsk., Celastraceae) is a perennial shrub that was introduced to Israel by Yemenite immigrants. Khat young leaves are chewed as a stimulant. The main stimulating active principles in this plant are the phenylpropylamino alkaloids (-)-cathinone [(S)-alpha-aminopropiophenone], (+)-cathine
Hesham Abdul Aziz et al.
The journal of sexual medicine, 6(3), 682-695 (2009-01-16)
Khat (Catha edulis) is an evergreen tree/shrub that is thought to affect sexual motivation or libido. Its positive effect on sexual desire is more frequently observed in females than in males and occurs when khat is chewed. Thus, khat's effects
Veniero Gambaro et al.
Forensic science international, 217(1-3), 87-92 (2011-10-26)
In the last years, all the vegetable material supposed to belong to the Catha Edulis species, seized at the Malpensa and Orio al Serio airports, were analyzed in our laboratory on behalf of the Tribunals of Busto Arsizio and Bergamo
Keep an eye on the pupil: developing countries under chemical attack.
Husni Al-Goshae et al.
Internal and emergency medicine, 4(3), 231-233 (2009-04-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service