13110
Magnesium
according to Grignard, ≥99.5%, turnings
About This Item
vapor pressure
1 mmHg ( 621 °C)
assay
≥99.5%
form
turnings
autoignition temp.
950 °F
quality
according to Grignard
reaction suitability
reagent type: reductant
resistivity
4.46 μΩ-cm, 20°C
impurities
≤0.01% insoluble in HCl
bp
1090 °C (lit.)
mp
648 °C (lit.)
density
1.74 g/mL at 25 °C (lit.)
cation traces
Fe: ≤500 mg/kg
SMILES string
[Mg]
InChI
1S/Mg
InChI key
FYYHWMGAXLPEAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service