Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13010

Sigma-Aldrich

4-Benzoyl-3-methyl-1-phenyl-2-pyrazolin-5-one

≥99.0% (GC)

Synonym(s):

1-Phenyl-3-methyl-4-benzoyl-2-pyrazolin-5-one, HPMBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O2
CAS Number:
Molecular Weight:
278.31
Beilstein/REAXYS Number:
888605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥99.0% (GC)

mp

90-92 °C (lit.)
90-92 °C

SMILES string

CC1=NN(c2ccccc2)C(=O)C1C(=O)c3ccccc3

Looking for similar products? Visit Product Comparison Guide

Other Notes

Complexing agent (pKa = 3.92) forming very stable metal complexes; used for the extraction and separation of trace metals for AAS[1]; Determination of metals by HPLC separation of complexes[2][3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Akama et al.
Analytical Sciences, 8, 41-41 (1992)
Y. Akama et al.
Journal of Chromatography A, 633, 129-129 (1993)
S. Umetami et al.
Analytical Chemistry, 64, 2288-2288 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service