Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12227

Sigma-Aldrich

3-(1,2,4-Triazol-1-yl)-L-alanine

Wacker Chemie AG, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)

Synonym(s):

(S)-2-Amino-3-(1,2,4-triazol-1-yl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4O2
CAS Number:
Molecular Weight:
156.14
Beilstein/REAXYS Number:
4181612
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

produced by Wacker Chemie AG, Burghausen, Germany

assay

≥95.0% (HPLC)

optical purity

ee: >99% (due to stereospecific enzymatic synthesis)

manufacturer/tradename

Wacker Chemie AG

storage temp.

2-8°C

SMILES string

N[C@@H](Cn1cncn1)C(O)=O

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service