Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

121223

Sigma-Aldrich

4,4′-Trimethylenebis(1-piperidineethanol)

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34N2O2
CAS Number:
Molecular Weight:
298.46
EC Number:
MDL number:
UNSPSC Code:
12162002

assay

98%

bp

270 °C/10 mmHg (lit.)

mp

94-96 °C (lit.)

SMILES string

OCCN1CCC(CCCC2CCN(CCO)CC2)CC1

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service