Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

119717

Sigma-Aldrich

Di-sec-butylphenol,mixture of isomers

technical grade, ≥85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C2H5CH(CH3)]2C6H3OH
CAS Number:
Molecular Weight:
206.32
EC Number:
UNSPSC Code:
12352103

grade

technical grade

vapor pressure

<0.01 mmHg ( 20 °C)

assay

≥85%

refractive index

n20/D 1.508 (lit.)

bp

255-260 °C (lit.)

mp

−42 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

260.6 °F - closed cup

flash_point_c

127 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service