113026
Glyceryl tributyrate
Synonym(s):
1,2,3-Tributyrylglycerol, Glycerol tributyrate, Tributyrin
About This Item
Recommended Products
assay
98%
refractive index
n20/D 1.435 (lit.)
bp
129-131 °C/0.03 mmHg (lit.)
287-288 °C (lit.)
solubility
H2O: insoluble
alcohol: very soluble
diethyl ether: very soluble
density
1.032 g/mL at 20 °C (lit.)
SMILES string
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC
InChI
1S/C15H26O6/c1-4-7-13(16)19-10-12(21-15(18)9-6-3)11-20-14(17)8-5-2/h12H,4-11H2,1-3H3
InChI key
UYXTWWCETRIEDR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
345.2 °F - closed cup
flash_point_c
174 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service