Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

112097

Sigma-Aldrich

Thymol

98%

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein/REAXYS Number:
1907135
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

1 mmHg ( 64 °C)

assay

98%

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junaid U Rehman et al.
Journal of medical entomology, 53(1), 157-165 (2015-11-22)
Mosquitoes are vectors for many pathogens resulting in many deaths of humans. Repellents play an important role in reducing mosquito bites and the spread of mosquito-borne diseases. Currently, Klun & Debboun (K & D) and human-arm-based bioassay systems are used
Lihe Yeo et al.
Food chemistry, 199, 393-400 (2016-01-19)
This study investigated how hydrophobicity, solubility and the concentration of flavour compounds related to inclusion complexation by dispersed native high amylose maize starch (HAMS). The effect of native lipid on flavour retention and the effect of time (one day to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service