10902
Isobutyl acetoacetate
produced by Wacker Chemie AG, Burghausen, Germany, ≥96.0% (GC)
Synonym(s):
ACIB
About This Item
Recommended Products
grade
produced by Wacker Chemie AG, Burghausen, Germany
assay
≥96.0% (GC)
manufacturer/tradename
Wacker Chemie AG
refractive index
n20/D 1.424 (lit.)
bp
100 °C/22 mmHg (lit.)
density
0.98 g/mL at 25 °C (lit.)
SMILES string
CC(C)COC(=O)CC(C)=O
InChI
1S/C8H14O3/c1-6(2)5-11-8(10)4-7(3)9/h6H,4-5H2,1-3H3
InChI key
ZYXNLVMBIHVDRH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
172.4 °F - closed cup
flash_point_c
78 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service