107697
(Dimethylamino)acetone
98%
Synonym(s):
1-Dimethylamino-2-propanone
About This Item
Recommended Products
assay
98%
refractive index
n20/D 1.414 (lit.)
bp
119-120 °C (lit.)
density
0.883 g/mL at 25 °C (lit.)
SMILES string
CN(C)CC(C)=O
InChI
1S/C5H11NO/c1-5(7)4-6(2)3/h4H2,1-3H3
InChI key
VFPKIWATTACVJR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
signalword
Warning
hcodes
pcodes
Hazard Classifications
Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
86.0 °F - closed cup
flash_point_c
30 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service