105562
4-Aminophenylmercuric acetate
technical grade
Synonym(s):
4-(Acetoxymercurio)aniline, APMA
About This Item
grade
technical grade
mp
163-165 °C (lit.)
SMILES string
CC(=O)O[Hg]c1ccc(N)cc1
InChI
1S/C6H6N.C2H4O2.Hg/c7-6-4-2-1-3-5-6;1-2(3)4;/h2-5H,7H2;1H3,(H,3,4);/q;;+1/p-1
InChI key
RXSUFCOOZSGWSW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service