Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

104833

Sigma-Aldrich

Trichloro(propyl)silane

98%

Synonym(s):

Dow Corning® product Z-1213, Propyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SiCl3
CAS Number:
Molecular Weight:
177.53
Beilstein/REAXYS Number:
1734990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.430 (lit.)

bp

123-124 °C (lit.)

density

1.195 g/mL at 25 °C (lit.)

SMILES string

CCC[Si](Cl)(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Legal Information

Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.
Dow Corning is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

87.8 °F - closed cup

flash_point_c

31 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service