Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

104183

Sigma-Aldrich

cis-4-Methylcyclohexanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.461 (lit.)

bp

172-173 °C (lit.)

density

0.917 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CC[C@@H](O)CC1

InChI

1S/C7H14O/c1-6-2-4-7(8)5-3-6/h6-8H,2-5H2,1H3/t6-,7+

InChI key

MQWCXKGKQLNYQG-KNVOCYPGSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kutsukake et al.
Gene, 245(1), 31-42 (2000-03-14)
We characterized molecular profiles of a new olfactory mutant line, honoka (hono), which was found among 500 viable P-element insertion lines screened first by 5-bromo-4-chloro-3-indrolyl-beta-D-galactopyranoside (X-gal) staining on the third segment of the antenna, and then by behavioral assays to
J R Linley
Medical and veterinary entomology, 3(4), 347-352 (1989-10-01)
Laboratory experiments tested the effects of p-cresol or 4-methylcyclohexanol at concentrations of 1, 10 and 50 ppm, on oviposition by the mosquitoes Toxorhynchites brevipalpis Theobald, Tx. amboinensis (Doleschall) and Tx. splendens (Wiedemann). A 5 + 5 ppm mixture of the
Jianzeng Guo et al.
Science (New York, N.Y.), 309(5732), 307-310 (2005-07-09)
Different modalities of sensation interact in a synergistic or antagonistic manner during sensory perception, but whether there is also interaction during memory acquisition is largely unknown. In Drosophila reinforcement learning, we found that conditioning with concurrent visual and olfactory cues
Studies of 4-methylcyclohexanol: an Aedes triseriatus (Diptera: Culicidae) oviposition attractant.
M D Bentley et al.
Journal of medical entomology, 19(5), 589-592 (1982-10-14)
Xianju Zhou et al.
Journal of biological rhythms, 20(3), 237-244 (2005-04-27)
Odors elicit a number of behavioral responses, including attraction and repulsion in Drosophila. In this study, the authors used a T-maze apparatus to show that wild-type Drosophila melanogaster exhibit a robust circadian rhythm in the olfactory attractive and repulsive responses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service