Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

102156

Supelco

Sulfonazo III sodium salt

Synonym(s):

2,7-Bis(2-sulfophenylazo)chromotropic acid tetrasodium salt, 3,6-Bis(2-sulfophenylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12N4Na4O14S4
CAS Number:
Molecular Weight:
776.57
Beilstein/REAXYS Number:
1900148
EC Number:
MDL number:
UNSPSC Code:
12352200

λmax

567 nm
629 nm (2nd)

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(\N=N\c2ccccc2S([O-])(=O)=O)c(cc3cc(c(\N=N\c4ccccc4S([O-])(=O)=O)c(O)c13)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O14S4.4Na/c27-21-18-11(9-16(43(35,36)37)19(21)25-23-12-5-1-3-7-14(12)41(29,30)31)10-17(44(38,39)40)20(22(18)28)26-24-13-6-2-4-8-15(13)42(32,33)34;;;;/h1-10,27-28H,(H,29,30,31)(H,32,33,34)(H,35,36,37)(H,38,39,40);;;;/q;4*+1/p-4/b25-23+,26-24+;;;;

InChI key

FCMRBZBGGXLKBC-KBJWUIGUSA-J

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service