Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

102148

Sigma-Aldrich

Safranine O

certified by the Biological Stain Commission, Dye content 95 %

Synonym(s):

3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Colour Index Number:
50240
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

product line

EMPROVE® Exp

composition

Dye content, 95%

λmax

530 nm

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Carrino et al.
Biochemistry international, 24(3), 485-495 (1991-06-01)
Recently, an assay for quantification of glycosaminoglycans has been reported based on precipitation with Safranin O (Lammi, M. and Tammi, M. (1988) Anal. Biochem. 168, 352-357). In this procedure, the precipitate which forms when the glycosaminoglycan or proteoglycan is mixed
P X Petit et al.
European journal of biochemistry, 194(2), 389-397 (1990-12-12)
Washed and purified rat- or mouse-liver mitochondria exhibiting high membrane integrity and metabolic activity were studied by flow cytometry. The electrophoretic accumulation/redistribution of cationic lipophilic probes, rhodamine 123, safranine O and a cyanine derivative, 3,3'-dihexyloxadicarbocyanine iodide, during the energization process
Jun Liu et al.
Scientific reports, 5, 17569-17569 (2015-12-03)
The development of the cornea, a highly specialized transparent tissue located at the anterior of the eye, is coordinated by a variety of molecules and cells. Here, we report that mast cells (MCs), recently found to be involved in morphogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service