07172
1-(4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecyl)-3,1-benzoxazine-2,4(1H)-dione
≥97.0%
Synonym(s):
1-(1H,1H,2H,2H,3H,3H-Perfluoroundecyl)-3,1-benzoxazine-2,4(1H)-dione, 1-[3-(Perfluorooctyl)propyl]-3,1-benzoxazine-2,4(1H)-dione
About This Item
assay
≥97.0%
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCN1C(=O)OC(=O)c2ccccc12
InChI
1S/C19H10F17NO3/c20-12(21,6-3-7-37-9-5-2-1-4-8(9)10(38)40-11(37)39)13(22,23)14(24,25)15(26,27)16(28,29)17(30,31)18(32,33)19(34,35)36/h1-2,4-5H,3,6-7H2
InChI key
JRKNOQQQWMFWME-UHFFFAOYSA-N
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
target_organs
Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service