All Photos(1)
About This Item
Linear Formula:
C2Cl5CHCl2
CAS Number:
Molecular Weight:
285.21
Beilstein/REAXYS Number:
1748917
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
99%
bp
165-166 °C/90 mmHg (lit.)
mp
29-30 °C (lit.)
SMILES string
ClC(Cl)C(Cl)(Cl)C(Cl)(Cl)Cl
InChI
1S/C3HCl7/c4-1(5)2(6,7)3(8,9)10/h1H
Inchi Key
YFIIENAGGCUHIQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Preparation of Aliphatic Fluorine Compounds.
Henne AL.
Organic Reactions, 2 (1944)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service