MilliporeSigma
All Photos(1)
Key Documents
W455320
Sigma-Aldrich
cis-3-Nonenyl acetate
≥95%, stabilized, FG
Sign Into View Organizational & Contract Pricing
Select a Size
W455320-SAMPLE-K
$52.50
100 G
$288.00
1 KG
$840.00
$52.50
In StockDetails
Request a Bulk Order
Select a Size
Change View
W455320-SAMPLE-K
$52.50
100 G
$288.00
1 KG
$840.00
About This Item
Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
4553
UNSPSC Code:
12164502
Flavis number:
09.672
NACRES:
NA.21
organoleptic:
green; herbaceous; pear; tropical
grade:
FG
Halal
Kosher
agency:
follows IFRA guidelines
food allergen:
no known allergens
$52.50
In StockDetails
Request a Bulk Order
grade
FG
Halal
Kosher
Quality Level
agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
assay
≥95%
contains
synthetic α-tocopherol as stabilizer
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
organoleptic
green; herbaceous; pear; tropical
SMILES string
O(CC\C=C/CCCCC)C(=O)C
InChI
1S/C11H20O2/c1-3-4-5-6-7-8-9-10-13-11(2)12/h7-8H,3-6,9-10H2,1-2H3/b8-7-
InChI key
LRQNQAVSERJKNU-FPLPWBNLSA-N
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Questions
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service