Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Hi-Flow
™
Plus Membrane 90
Products
Product Category
membranes (10)
rolls (6)
sheets (4)
Brand
Millipore (10)
Length (cm)
Length (m)
Quality Segment
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
HF09004XSS
Hi-Flow
™
Plus Membrane 90,
backing thickness 4 mil (100 μm), filter W × L 30.5 cm × 25 cm, pack of 5 sheets
Expand
View Pricing
HF09002XSS
Hi-Flow
™
Plus Membrane 90,
backing thickness 2 mil (50 μm), filter W × L 30.5 cm × 25 cm, pack of 5 sheets
Expand
View Pricing
HF090MC100
Hi-Flow
™
Plus Membrane 90,
card W × L 6 cm × 30.1 cm, pack of 100 sheets
Expand
View Pricing
HF090MC5PK
Hi-Flow
™
Plus Membrane 90,
pack of 5 laminated membrane cards, size 6 cm × 30.1 cm
Expand
View Pricing
SHF0900425
Hi-Flow
™
Plus Membrane 90,
backing thickness 4 mil, width 25 mm, pack of 1 slit roll
Expand
View Pricing
BPCKHF09004S25
Hi-Flow
™
Plus Membrane 90,
backing thickness 4 mil (100 μm), slit width 25 mm, pack of 11 roll
Expand
View Pricing
SHF0900225
Hi-Flow
™
Plus Membrane 90,
backing thickness 2 mil, width 25 mm, pack of 1 slit roll
Expand
View Pricing
BPCKHF09004S20
Hi-Flow
™
Plus Membrane 90,
backing thickness 4 mil (100 μm), slit width 20 mm, pack of 14 roll
Expand
View Pricing
BPCKHF09002S20
Hi-Flow
™
Plus Membrane 90,
backing thickness 2 mil (50 μm), slit width 20 mm, pack of 14 roll
Expand
View Pricing
BPCKHF09002S25
Hi-Flow
™
Plus Membrane 90,
backing thickness 2 mil (50 μm), slit width 25 mm, pack of 11 roll
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More