Enter the New Era of Media Preparation
Dispense up to 100 L of preheated, sterile & ISO compliant culture media with this bag and our new ReadyStream® System!
ON-DEMAND CULTURE MEDIA PREPARATION
Welcome to the future of media preparation with the ReadyStream® system. This innovative solution transforms traditional media preparation into a streamlined, efficient process that saves time, reduces contamination risks, and optimizes laboratory workflows.
The ReadyStream® system simplifies culture media preparation by allowing users to prepare and dispense sterile, preheated media on demand with just the push of a button. Gone are the days of labor-intensive, multi-step procedures involving cleaning, autoclaving, and manual handling of dehydrated media. Instead, our system enables the preparation of up to 100 liters of media in under 20 minutes, ensuring that your laboratory can operate at peak efficiency.
With the ReadyStream®system:
- No handling of dehydrated media
- No autoclaving
- No media kitchen needed
- No bottles, no washing
- Small footprint
- Less variability of media quality
- Media readily heated for use
- Up to 100 L of ready-to-use culture media, and nothing you can’t carry
The ReadyStream® system reinvents media preparation and dispensing, the core principle is that the ReadyStream® system dilutes concentrated media with sterile water. This has huge advantages.
Watch the video showcasing the ReadyStream® system, which offers an innovative method for preparing microbiological culture media. By simplifying media handling, the ReadyStream® system enhances laboratory workflows and streamlines the preparation and dispensing of culture media.
The main advantages of the ReadyStream® system
10x Media Concentrate at the Ready
The ReadyStream® system offers on-demand, concentrated media that gets diluted with sterile water to the correct concentration and dispense media into your sample bag.
A 3-liter ReadyStream® media bag lets you dispense up to 30 liters of culture media and a 10-liter bag up to 100 liters. The 10x concentrated media can be maintained for up to 5 days.
No Need to Autoclave Media
To create the concentrate, the water is sterilized by the ReadyStream® filter set before entering the ReadyStream® media bag containing the irradiated and ISO-compliant GranuCult® dehydrated culture media. After this step, which takes 20 minutes, the concentrate will allow you to prepare up to 100 liters of media on demand.
Instant Preheated Culture Media
Any time you need the ReadyStream® system dispenses pre-heated culture media at a preset temperature of between 30 and 42 °C and a volume of between 90 to 9999 mL.
Connect the ReadyStream® system to any dilutor through an intermediate bottle serving as media reservoir. This allows multiple setups for greater flexibility, e.g. transportation of the bottle within the lab and connection to multiple dilutors. Also, it can facilitate the addition of supplements.
Watch the Webinar on the ReadyStream® System
Watch our on-demand webinar on ReadyStream® system, which showcases the live demo of the system and how to use it. Also, see our experts answer some of the questions raised in the session. However, you can still share your questions just register to our platform to get access to the webinar and simply submit your questions.
Network error: Failed to fetch
Discover the ReadyStream® system for effortless media preparation. Simplify your workflow with on-demand, sterile media—no autoclaving or heavy lifting required.
A comparative study on time, energy and water requirements, performed by an independent food QC laboratory.
The ReadyStream® filter set is a sterilizing filter for deionized water to be used together with ReadyStream® media bags on the ReadyStream® system.
The seamless connection between ReadyStream® and Milli-Q® systems can support your lab to integrate preparation workflows and maximize space.
Dispensing Media and Filling Sample Bags on the ReadyStream® System
For the preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter, from food and animal feed, water and other materials.
For the preliminary non-selective enrichment of bacteria, especially Salmonella, from food and other materials.
The self-generated test data matrixes can be integrated into a standard operating test procedure or printed and attached to a product to be tested.